Assay Details
| Gene Symbol: C130046K22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_102388.1 | 3 | 122 | |||
| NR_102389.1 | 3 | 122 | ||||
| GenBank mRNA | AK142001.1 | - | 3 | 122 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | golgi SNAP receptor complex member 2 | Gosr2 |
2310032N09Rik, C76855, Gs27, SNARE, membrin |
-25.0kb | Anti-sense | Chr. 11 - 103676849 - 103697738 on NCBI Build GRCm38 |
| Downstream |
wingless-type MMTV integration site family, member 9B |
Wnt9b |
Wnt14b, Wnt15, clf, clf1, wnt-14b, wnt-15 |
1.8kb | Anti-sense | Chr. 11 - 103727363 - 103749821 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

C130046K22Rik
Genomic Map