Assay Details
| Gene Symbol: 1700020G03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK006158.1 | - | 1 | 850 | 84 | |
| BB806657.1 | - | 1 | 322 | 84 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | transmembrane protein 59-like | Tmem59l | 5330410G16Rik | -713.0 | Anti-sense | Chr. 8 - 70483863 - 70487468 on NCBI Build GRCm38 |
| Downstream | cytokine receptor-like factor 1 | Crlf1 |
CLF-1, CRLM3, NR6.1 |
4.0kb | Sense | Chr. 8 - 70493107 - 70504081 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

1700020G03Rik
Genomic Map