Assay Details
| Gene Symbol: Golga7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042484.1 | NP_001035949.1 | 6 | 1153 | 89 | |
| NM_020585.2 | NP_065610.2 | 5 | 1366 | 89 | ||
| GenBank mRNA | AB041568.1 | - | 5 | 1302 | 89 | |
| AK048175.1 | - | 6 | 1129 | 89 | ||
| AK077582.1 | - | 6 | 1127 | 89 | ||
| AK150462.1 | - | 6 | 1147 | 89 | ||
| AK150696.1 | - | 5 | 1308 | 89 | ||
| AK151353.1 | - | 6 | 1147 | 89 | ||
| AK152520.1 | - | 6 | 1147 | 89 | ||
| AK152606.1 | - | 6 | 1126 | 89 | ||
| AK153213.1 | - | 6 | 1147 | 89 | ||
| AK158227.1 | - | 5 | 1272 | 89 | ||
| AK170129.1 | - | 6 | 1132 | 89 | ||
| AK211526.1 | - | 1 | 183 | 89 | ||
| AK212515.1 | - | 1 | 183 | 89 | ||
| BC016894.1 | - | 6 | 1105 | 4223937 | 89 | |
| BC019697.1 | - | 5 | 1291 | 89 | ||
| BC058523.1 | - | 5 | 1209 | 5717934 | 89 | |
| BC093518.1 | - | 6 | 1123 | 3157520 | 89 | |
| BC103796.1 | - | 6 | 1118 | 6519018 | 89 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25715749 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25715748 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25715745 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25715751 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2120682 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2027323 | BC103796.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2027323 | BC019697.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2027323 | BC058523.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2027323 | BC093518.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2027323 | AK158227.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 303475 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 115248 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 42625 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 742390 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 871819 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 626490 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 929083 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 816519 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 53863 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 65330 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 166602 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P110767 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P174903 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2027323 | BC103796.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2027323 | BC019697.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2027323 | BC058523.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2027323 | BC093518.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2027323 | AK158227.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2120682 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25715732 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25715733 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25715745 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25715768 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25715731 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25715736 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25715766 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25715755 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25715758 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25715741 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25715737 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25715740 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25715729 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25715749 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25715738 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25715764 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25715742 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25715748 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25715762 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25715769 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25715757 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25715760 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25715727 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25715752 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P110767 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P174903 | - | - |
| Whole Mouse Genome G4122A | A_52_P398018 | AK089856.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P478942 | BC103796.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P478942 | BC019697.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P478942 | BC058523.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P478942 | BC093518.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P478942 | AK158227.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P130901 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P110767 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P174903 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06943717 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06943715 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06943719 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06943710 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06943713 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25715751 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25715748 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25715745 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25715749 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25715769 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25715673 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25715762 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25715692 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25715581 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25715766 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25715615 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25715576 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25715786 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25715773 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25715675 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25715676 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25715647 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25715697 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25715718 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25715671 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25715657 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25715679 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25715752 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25715691 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25715589 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25715705 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25715649 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25715584 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25715582 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25715596 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25715640 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25715654 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25715578 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25715787 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25715651 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25715602 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25715600 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25715748 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25715741 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25715669 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25715695 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25715775 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25715708 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25715755 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25715646 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25715715 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25715777 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25715588 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25715585 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25715765 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25715665 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25715611 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25715716 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25715683 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25715662 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25715693 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25715686 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25715736 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25715710 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25715608 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25715677 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25715660 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25715614 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25715680 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25715613 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25715618 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25715621 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25715696 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25715729 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25715690 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25715732 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25715742 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25715738 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25715643 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25715703 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25715781 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25715779 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25715652 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25715593 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25715612 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25715714 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25715772 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25715575 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25715776 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25715733 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25715749 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25715641 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25715727 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25715637 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25715655 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25715731 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25715766 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25715765 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25715644 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25715696 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25715738 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07306074 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25715768 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25715762 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25715733 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25715654 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07317787 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07339202 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07420354 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07085500 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06842984 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25715640 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07420355 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25715708 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25715652 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06943710 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25715695 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25715714 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25715712 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25715715 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25715655 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25715775 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25715729 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25715703 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07331195 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25715641 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25715705 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07133824 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25715776 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25715782 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25715643 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25715757 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25715741 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25715637 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06954907 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25715773 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25715769 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25715777 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25715742 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25715701 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25715749 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25715718 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06768719 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25715719 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25715709 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25715772 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25715710 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06943717 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25715748 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25715697 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25715716 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25715779 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07373722 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25715725 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25715752 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25715755 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25715649 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25715727 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25715647 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10577640 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10577636 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10577638 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10577637 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10577634 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10577639 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10532309 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10577635 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1415672_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10577633 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10532308 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10570955 | - | - |
| Mouse Genome 430 2.0 Array | 1415672_at | - | - |
| Mouse Genome 430 2.0 Array | 1415672_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 57437_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 57437_at | - | - |
| Mouse Expression Set 430 | 1415672_at | - | - |
| Mouse Expression Set 430 | 1415672_PM_at | - | - |
| Mouse Expression Set 430 | 1415672_at | - | - |
| Mouse Expression Set 430 | 1415672_PM_at | - | - |
| Murine Genome U74v2 Set | 93256_PM_at | - | - |
| Murine Genome U74v2 Set | 93256_at | - | - |
| Mouse Genome 430A 2.0 Array | 1415672_at | - | - |
| Mouse Genome 430A 2.0 Array | 1415672_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1415672_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1415672_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1415672_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1415672_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6980121 | - | - |
| MouseWG-6 v2 Expression BeadChip | 106760687 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 106760687 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6980121 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006980121 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2666519 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1254148 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2666519 | - | - |
| mouse-6 v1.1 (Illumina) | scl057437.1_41-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6980121 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 106760687 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl057437.1_41-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 106760687 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6980121 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3157764 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2666519 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2666519 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3157764 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1254148 | - | - |

Golga7
Genomic Map