Assay Details
| Gene Symbol: Slc8a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_104580.1 | 4 | 2072 | 79 | ||
| GenBank mRNA | AF108396.1 | - | 1 | 70 | 79 | |
| AF108397.1 | - | 1 | 70 | 79 | ||
| AF108398.1 | - | 1 | 70 | 79 | ||
| DQ388998.1 | - | 3 | 1962 | 79 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Slc8a1
Genomic Map