Assay Details
| Gene Symbol: Armcx4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202500.3 | NP_001189429.2 | 6 | 7818 | 126 | |
| GenBank mRNA | BC025879.1 | - | 1 | 1187 | 126 | |
| BC030323.1 | - | 2 | 3152 | 126 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Armcx4
Genomic Map