Assay Details
| Gene Symbol: Sec22a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK037540.1 | - | 2 | 617 | 85 | |
| BC057176.1 | - | 1 | 595 | 85 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Sec22a
Genomic Map