Assay Details
| Gene Symbol: E230006M18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK053951.1 | - | 1 | 3940 | 85 | |
| BB548263.2 | - | 1 | 645 | 85 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

E230006M18Rik
Genomic Map