Assay Details
| Gene Symbol: D030041H20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK083531.1 | - | 1 | 54 | 93 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 3075 | Mir3075 |
mir-3075, mmu-mir-3075 |
-13.9kb | Sense | Chr. 14 - 25534439 - 25534523 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 4931406H21 gene | 4931406H21Rik | 37.7kb | Sense | Chr. 14 - 25586804 - 25590661 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

D030041H20Rik
Genomic Map