Assay Details
| Gene Symbol: Gm30414 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK036865.1 | - | 3 | 2434 | 115 | |
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | BB143266.2 | - | 589 | Chr. - - [-] on Build | 115 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 26901 |
Gm26901 | -117.5kb | Anti-sense | Chr. 1 - 7314638 - 7398666 on NCBI Build GRCm38 | |
| Downstream |
syntrophin, gamma 1 |
Sntg1 |
4933426D16Rik, G1SYN, SYN4 |
841.6kb | Anti-sense | Chr. 1 - 8359739 - 9301061 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gm30414
Genomic Map