Assay Details
| Gene Symbol: Prl2c1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045532.2 | NP_001038997.1 | 5 | 660 | 127 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2183208 | NM_001045532.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P103122 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2183208 | NM_001045532.1 | mouse_transcriptome |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10619019 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903790 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903771 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903750 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903772 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903761 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903779 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903848 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903748 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903760 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903783 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903759 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903751 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903765 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903849 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903773 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903781 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903855 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903770 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903852 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903789 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903758 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903843 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903788 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903853 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903791 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903778 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903776 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903766 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903767 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903837 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903756 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903832 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903786 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903763 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903745 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28903781 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28903760 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10858163 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28903765 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28903796 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28903778 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10747182 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28903770 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28903759 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28903751 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28903764 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28903779 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28903750 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28903786 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28903763 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28903767 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28903771 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28903784 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28903791 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28903788 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28903790 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28903776 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28903773 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28903783 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28903761 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28903758 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28903789 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10619019 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28903756 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28903807 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10404332 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10404335 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10404334 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10404333 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10404330 | - | - |
| Affymetrix Mouse Clariom S Array | PSR1300001897.mm.1 | NM_001045532.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300001897.mm.1 | TC1300000305.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1300001901.mm.1 | NM_001045532.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300001901.mm.1 | TC1300000305.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1300001895.mm.1 | NM_001045532.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300001895.mm.1 | TC1300000305.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1300001900.mm.1 | NM_001045532.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300001900.mm.1 | TC1300000305.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1300001904.mm.1 | NM_001045532.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300001904.mm.1 | TC1300000305.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1300001899.mm.1 | NM_001045532.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300001899.mm.1 | TC1300000305.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1300001902.mm.1 | NM_001045532.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300001902.mm.1 | TC1300000305.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1437515_PM_at | - | - |
| Mouse Expression Set 430 | 1437515_at | - | - |
| Murine Genome U74v2 Set | 161590_r_at | - | - |
| Murine Genome U74v2 Set | 161590_PM_r_at | - | - |
| Murine Genome U74v2 Set | 94838_r_at | - | - |
| Murine Genome U74v2 Set | 94838_PM_r_at | - | - |
| Murine Genome U74v2 Set | 93883_PM_at | - | - |
| Murine Genome U74v2 Set | 93883_at | - | - |
| Mouse Genome 430A 2.0 Array | 1437515_at | - | - |
| Mouse Genome 430A 2.0 Array | 1437515_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseRef-8 v2 Expression BeadChip | ILMN_2826278 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2826278 | - | - |

Prl2c1
Genomic Map