Assay Details
| Gene Symbol: Ccnb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK136259.1 | - | 9 | 2249 | 113 | |
| BC080202.1 | - | 8 | 2022 | 3603637 | 113 | |
| X64713.1 | - | 9 | 2220 | 113 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ccnb1
Genomic Map