Assay Details
| Gene Symbol: Prlr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001253781.1 | NP_001240710.1 | 10 | 1422 | 109 | |
| GenBank mRNA | AB643813.1 | - | 8 | 1191 | 109 | |
| AK050317.1 | - | 11 | 1565 | 109 | ||
| AK052679.1 | - | 10 | 1320 | 109 | ||
| AK054381.1 | - | 11 | 1936 | 109 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Prlr
Genomic Map