Assay Details
| Gene Symbol: 1700122D07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK007236.1 | - | 2 | 345 | 81 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | frizzled homolog 7 (Drosophila) | Fzd7 | Fz7 | -47.9kb | Anti-sense | Chr. 1 - 59482147 - 59486955 on NCBI Build GRCm38 |
| Downstream | SMT3 suppressor of mif two 3 homolog 1 (yeast) | Sumo1 |
GMP1, PIC1, SENTRIN, SMT3, SMT3H3, SMTP3, SUMO-1, Smt3C, Ubl1 |
104.4kb | Sense | Chr. 1 - 59639434 - 59670834 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

1700122D07Rik
Genomic Map