Assay Details
| Gene Symbol: Gm35585 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK035376.1 | - | 2 | 1329 | 112 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | adaptor-related protein complex 1 associated regulatory protein | Ap1ar |
AA407621, C78909, Gadkin |
-26.2kb | Anti-sense | Chr. 3 - 127807007 - 127837518 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 5730508B09 gene | 5730508B09Rik |
2700063A17Rik, C80506 |
4.4kb | Anti-sense | Chr. 3 - 127869688 - 127896323 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gm35585
Genomic Map