Assay Details
| Gene Symbol: 6330418B08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK018197.1 | - | 1 | 2443 | 166 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | regulator of G protein signaling 7 | Rgs7 | Sense | Chr. 1 - 175059076 - 175493238 on NCBI Build GRCm38 | ||
| Downstream | fumarate hydratase 1 | Fh1 |
Fh, Fh-1 |
536.5kb | Sense | Chr. 1 - 175601378 - 175625635 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

6330418B08Rik
Genomic Map