Assay Details
| Gene Symbol: A330095P18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK039723.1 | - | 1 | 1256 | 137 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | fibrosin-like 1 | Fbrsl1 |
2410025L10Rik, AI467568, Gm1049, mKIAA1545 |
Sense | Chr. 5 - 110361751 - 110449233 on NCBI Build GRCm38 | |
| Downstream |
predicted gene, 29766 |
Gm29766 | 59.7kb | Sense | Chr. 5 - 110429875 - 110431955 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Agilent
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 258246 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 239384 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 414226 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 450610 | AK084495.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 450610 | AK141859.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 866641 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 805154 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 702962 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 509052 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 195976 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 78581 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 78934 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 318777 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 596538 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 712334 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 247065 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 610461 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 96061 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 676977 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 756625 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 344440 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 749429 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 503127 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 918523 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 430849 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 570231 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 631793 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 698632 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 44674 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 543576 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 105424 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 871916 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 493025 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 141208 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 570649 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 28556 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 731781 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 262367 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 935150 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 596478 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 297867 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 788862 | AK035450.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 275651 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 727613 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 128396 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 121090 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 178094 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 62255 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 177517 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 51577 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 5354 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 126364 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 111418 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P175489 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P143562 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01185769 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05693442 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P143562 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05693650 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05693546 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05693569 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01185864 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05693621 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05693578 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01185798 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01185782 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05693631 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P05693631 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P01185769 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P01185798 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P143562 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23979930 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23980069 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04381131 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04513017 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23980318 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23979935 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23979933 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23980312 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23980118 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23980070 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23980072 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04108347 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04867244 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04513011 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04513034 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23980075 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04269547 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23979936 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23979939 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23980305 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23979937 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23979944 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23980073 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23980315 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04513061 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23980133 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23980074 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23980067 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23980313 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23979941 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23980308 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23980124 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23980137 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23979938 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23979940 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04690867 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23980314 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23980117 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23980132 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23980127 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04690865 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23980065 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23980078 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04269550 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04513056 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 869239 | - | - |

A330095P18Rik
Genomic Map