Assay Details
| Gene Symbol: Spaca6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK035632.1 | - | 1 | 1294 | 111 | |
| AK048874.1 | - | 1 | 1296 | 111 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Spaca6
Genomic Map