Assay Details
Gene Symbol: BC049715 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK147870.1 | - | 1 | 182 |
Gene Symbol: Smco3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039558.2 | NP_001034647.2 | 2 | 1866 | 182 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2162825 | AK047766.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2162825 | NM_001039558.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2162825 | AK045581.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2162825 | AK147870.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2078325 | NM_001039558.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2078325 | AK045581.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2078325 | AK147870.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P160112 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2078325 | NM_001039558.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2078325 | AK045581.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2078325 | AK147870.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2162825 | AK047766.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2162825 | NM_001039558.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2162825 | AK045581.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2162825 | AK147870.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24874666 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24874675 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24874678 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24874723 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24874712 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24874681 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24874722 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24874711 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24874696 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24874694 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24874716 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24874707 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24874693 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24874686 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24874706 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24874725 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24874671 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24874727 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24874700 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24874729 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24874697 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24874673 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24874688 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24874714 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24874702 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24874698 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24874677 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24874704 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24874740 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24874730 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24874705 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24874718 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24874674 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24874734 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24874690 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24874708 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24874710 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24874684 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24874701 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24874679 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24874736 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24874735 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24874738 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24874732 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24874719 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24874668 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24874676 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24874670 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24874699 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24874692 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P117016 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874732 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874702 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874670 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874678 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874644 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874655 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874690 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874697 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874716 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874659 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874653 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874647 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874684 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874629 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874718 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874663 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874693 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874598 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874706 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874612 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874632 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874642 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874633 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874673 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874607 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874671 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874648 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874640 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874692 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874590 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874608 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874603 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874601 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874727 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874619 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874675 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874685 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874734 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874699 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874701 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874630 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874635 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874708 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874711 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874714 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874638 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874668 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874736 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874622 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874729 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874592 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874694 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874660 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874625 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874616 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874735 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874657 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874597 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874725 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874666 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874710 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874662 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874704 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874595 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874600 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874676 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874722 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874696 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874677 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874623 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874738 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874700 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874627 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874650 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24874606 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24874732 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24874730 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24874696 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24874719 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24874700 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24874725 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05679402 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24874710 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24874687 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05630589 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05674794 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05031875 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24874718 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24874686 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05679400 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05222382 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24874729 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24874706 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24874702 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24874722 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24874684 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05674791 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24874716 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24874685 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24874699 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24874701 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05222383 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05222364 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24874704 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24874708 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24874692 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24874690 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05222366 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24874736 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24874731 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10542404 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10542403 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10548872 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10548873 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10548874 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1457323_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10548871 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10542401 | - | - |
Mouse Genome 430 2.0 Array | 1457323_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1457323_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 654818_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 654818_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0600026159.mm.1 | TC0600003333.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0600026159.mm.1 | NM_001039558.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600026159.mm.1 | NM_001039558.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600026156.mm.1 | TC0600003333.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0600026156.mm.1 | NM_001039558.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600026156.mm.1 | NM_001039558.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600026157.mm.1 | TC0600003333.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0600026157.mm.1 | NM_001039558.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600026157.mm.1 | NM_001039558.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600026155.mm.1 | TC0600003333.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0600026155.mm.1 | NM_001039558.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600026155.mm.1 | NM_001039558.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600026158.mm.1 | TC0600003333.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0600026158.mm.1 | NM_001039558.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600026158.mm.1 | NM_001039558.2 | Refseq |
Mouse Expression Set 430 | 1457323_PM_at | - | - |
Mouse Expression Set 430 | 1457323_at | - | - |
Murine Genome U74v2 Set | 138541_PM_r_at | - | - |
Murine Genome U74v2 Set | 138541_r_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1457323_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1457323_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 402827 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 106420019 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 103170168 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 106420019 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 101050546 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2555588 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1243953 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1234867 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 103170168 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 106420019 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 101050546 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 106420019 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 103170168 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 101050546 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2801171 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2555705 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2555588 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2581500 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2801171 | - | - |
