| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0500021464.mm.1 |
TC0500002343.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500021464.mm.1 |
NM_024236.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500021464.mm.1 |
NM_024236.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500021468.mm.1 |
TC0500002343.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500021468.mm.1 |
NM_024236.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500021468.mm.1 |
NM_024236.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500021465.mm.1 |
TC0500002343.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500021465.mm.1 |
NM_024236.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500021465.mm.1 |
NM_024236.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500021461.mm.1 |
TC0500002343.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500021461.mm.1 |
NM_024236.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500021461.mm.1 |
NM_024236.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500021466.mm.1 |
TC0500002343.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500021466.mm.1 |
NM_024236.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500021466.mm.1 |
NM_024236.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500021467.mm.1 |
TC0500002343.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500021467.mm.1 |
NM_024236.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500021467.mm.1 |
NM_024236.2 |
Refseq |