Assay Details
| Gene Symbol: Gys1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030678.3 | NP_109603.2 | 16 | 3538 | 148 | |
| GenBank mRNA | AK085476.1 | - | 12 | 149 | ||
| AK169344.1 | - | 16 | 3551 | 165 | ||
| AK170404.1 | - | 16 | 3541 | 148 | ||
| AK171148.1 | - | 16 | 3510 | 165 | ||
| AK171426.1 | - | 16 | 3519 | 148 | ||
| X94616.1 | - | 16 | 3400 | 165 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gys1
Genomic Map