Assay Details
| Gene Symbol: Apold1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109914.1 | NP_001103384.1 | 2 | 2251 | 160 | |
| GenBank mRNA | AK085884.1 | - | 1 | 1079 | 160 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2062108 | NM_001109914.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2062108 | AK085884.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P109183 | NM_001109914.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P109183 | AK085884.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 189046 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 528202 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 128864 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 449832 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P109183 | NM_001109914.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P109183 | AK085884.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2062108 | NM_001109914.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2062108 | AK085884.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P109183 | NM_001109914.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P109183 | AK085884.1 | mouse_transcriptome |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24864560 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24864578 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24864505 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24864543 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24864605 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24864601 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24864586 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24864575 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24864490 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24864532 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24864617 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24864565 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24864549 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24864528 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24864599 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24864541 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24864611 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24864577 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24864529 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24864552 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24864551 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24864593 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24864510 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24864598 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24864513 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24864603 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24864544 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24864498 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24864489 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24864533 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24864506 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24864534 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24864597 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24864591 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24864608 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24864542 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24864521 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24864511 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24864508 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24864494 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24864589 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24864540 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24864584 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24864537 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24864522 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24864535 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24864547 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24864554 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24864558 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24864600 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24864500 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24864610 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24864596 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24864568 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24864580 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24864566 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05284387 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24864498 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24864549 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24864535 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05198465 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05163877 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05284412 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24864558 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24864553 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24864494 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05769497 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24864522 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05242953 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24864534 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24864533 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24864504 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05713420 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05301130 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24864529 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05205379 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05769498 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24864489 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24864513 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24864551 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24864552 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05294422 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24864547 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24864532 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24864524 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24864506 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24864565 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24864495 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24864573 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05284379 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24864571 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24864572 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24864566 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24864510 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05803597 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05301127 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24864523 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24864511 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24864526 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24864580 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24864557 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24864561 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05769495 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24864505 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24864568 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24864563 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24864574 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24864578 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10542320 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1441228_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10542319 | - | - |
| Mouse Genome 430 2.0 Array | 1441228_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1441228_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 381823_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 381823_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0600012791.mm.1 | TC0600001600.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600012791.mm.1 | NM_001109914.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600012791.mm.1 | NM_001109914.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600012789.mm.1 | TC0600001600.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600012789.mm.1 | NM_001109914.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600012789.mm.1 | NM_001109914.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600012790.mm.1 | TC0600001600.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600012790.mm.1 | NM_001109914.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600012790.mm.1 | NM_001109914.1 | Refseq |
| Mouse Expression Set 430 | 1441228_PM_at | - | - |
| Mouse Expression Set 430 | 1441228_at | - | - |
| Murine Genome U74v2 Set | 166870_PM_at | - | - |
| Murine Genome U74v2 Set | 166870_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1441228_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1441228_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 493818 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1222453 | - | - |

Apold1
Genomic Map