Assay Details
| Gene Symbol: Kbtbd6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034882.3 | NP_001030054.3 | 1 | 2037 | 103 | |
| GenBank mRNA | AK159684.1 | - | 1 | 2038 | 103 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29751456 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29751454 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2018904 | NM_001034882.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2018904 | AK159684.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 315362 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2018904 | NM_001034882.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2018904 | AK159684.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08175527 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P08175527 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29751456 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29751454 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751476 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751447 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751433 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751438 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751469 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751456 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751449 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751471 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751424 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751429 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751474 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751467 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751453 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751454 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751420 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751432 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751440 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751481 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751427 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751415 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751452 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751444 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751436 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751466 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751411 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751413 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29751418 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29751439 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29751449 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29751427 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11126469 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29751422 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29751452 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11752549 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29751443 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29751474 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29751453 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29751434 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29751432 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29751436 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29751477 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29751424 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11590810 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29751471 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29751444 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29751482 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29751438 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29751447 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29751465 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29751476 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29751433 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29751440 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29751437 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29751469 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29751479 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29751466 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29751481 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29751467 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10416656 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1439865_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1437130_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10416655 | - | - |
| Mouse Genome 430 2.0 Array | 1439865_at | - | - |
| Mouse Genome 430 2.0 Array | 1439865_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1437130_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1437130_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 432879_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 432879_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1400008816.mm.1 | NM_001034882.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400008816.mm.1 | NM_001034882.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400008816.mm.1 | TC1400001104.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1437130_at | - | - |
| Mouse Expression Set 430 | 1437130_PM_at | - | - |
| Mouse Expression Set 430 | 1439865_PM_at | - | - |
| Mouse Expression Set 430 | 1439865_at | - | - |
| Murine Genome U74v2 Set | 168250_f_at | - | - |
| Murine Genome U74v2 Set | 168250_PM_f_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1439865_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1437130_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1439865_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1437130_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_3161906 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_3161906 | - | - |
| mouse-6 v1.1 (Illumina) | GI_85702225-S | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3161906 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3161906 | - | - |

Kbtbd6
Genomic Map