Assay Details
| Gene Symbol: Fbll1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004147.3 | NP_001004147.1 | 1 | 1350 | 178 | |
| GenBank mRNA | AK140117.1 | - | 1 | 1056 | 178 | |
| BC052068.1 | - | 1 | 1040 | 5710336 | 178 | |
| BU701095.1 | - | 1 | 178 | |||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
box C/D snoRNA 3'-end processing
rRNA processing
tRNA processing
rRNA methylation
methylation
histone glutamine methylation
rRNA (adenine-N6,N6-)-dimethyltransferase activity
RNA binding
protein C-terminal carboxyl O-methyltransferase activity
tRNA (guanine-N2-)-methyltransferase activity
methyltransferase activity
C-methyltransferase activity
N-methyltransferase activity
O-methyltransferase activity
S-methyltransferase activity
RNA methyltransferase activity
mRNA methyltransferase activity
tRNA methyltransferase activity
protein methyltransferase activity
2-polyprenyl-6-methoxy-1,4-benzoquinone methyltransferase activity
rRNA methyltransferase activity
rRNA (uridine-2'-O-)-methyltransferase activity
rRNA (adenine-N6-)-methyltransferase activity
DNA-methyltransferase activity
tRNA (guanine-N1-)-methyltransferase activity
rRNA (cytosine-C5-)-methyltransferase activity
selenocysteine methyltransferase activity
arginine N-methyltransferase activity
protein-arginine N-methyltransferase activity
lysine N-methyltransferase activity
protein-lysine N-methyltransferase activity
tRNA (uracil) methyltransferase activity
tRNA (guanine) methyltransferase activity
tRNA (adenine) methyltransferase activity
tRNA (cytosine) methyltransferase activity
tRNA (cytosine-5-)-methyltransferase activity
tRNA (adenine-N1-)-methyltransferase activity
rRNA (adenine) methyltransferase activity
rRNA (cytosine) methyltransferase activity
rRNA (guanine) methyltransferase activity
rRNA (uridine) methyltransferase activity
transferase activity
protein C-terminal leucine carboxyl O-methyltransferase activity
1-phenanthrol methyltransferase activity
protein-arginine N5-methyltransferase activity
methylarsonite methyltransferase activity
dimethylarsinite methyltransferase activity
4,5-dihydroxybenzo(a)pyrene methyltransferase activity
1-hydroxypyrene methyltransferase activity
1-hydroxy-6-methoxypyrene methyltransferase activity
demethylmenaquinone methyltransferase activity
cobalt-precorrin-6B C5-methyltransferase activity
cobalt-precorrin-7 C15-methyltransferase activity
cobalt-precorrin-5B C1-methyltransferase activity
cobalt-precorrin-3 C17-methyltransferase activity
dimethylamine methyltransferase activity
hydroxyneurosporene-O-methyltransferase activity
tRNA (adenine-57, 58-N(1)-) methyltransferase activity
methylamine-specific methylcobalamin:coenzyme M methyltransferase activity
trimethylamine methyltransferase activity
methanol-specific methylcobalamin:coenzyme M methyltransferase activity
monomethylamine methyltransferase activity
P-methyltransferase activity
Se-methyltransferase activity
2-phytyl-1,4-naphthoquinone methyltransferase activity
tRNA (uracil-2'-O-)-methyltransferase activity
tRNA (cytosine-2'-O-)-methyltransferase activity
phosphomethylethanolamine N-methyltransferase activity
tRNA (cytosine-3-)-methyltransferase activity
rRNA (cytosine-2'-O-)-methyltransferase activity
rRNA (cytosine-N4-)-methyltransferase activity
trihydroxyferuloyl spermidine O-methyltransferase activity
histone-glutamine methyltransferase activity
box C/D snoRNA 3'-end processing
rRNA processing
tRNA processing
rRNA methylation
methylation
histone glutamine methylation
rRNA (adenine-N6,N6-)-dimethyltransferase activity
RNA binding
protein C-terminal carboxyl O-methyltransferase activity
tRNA (guanine-N2-)-methyltransferase activity
methyltransferase activity
C-methyltransferase activity
N-methyltransferase activity
O-methyltransferase activity
S-methyltransferase activity
RNA methyltransferase activity
mRNA methyltransferase activity
tRNA methyltransferase activity
protein methyltransferase activity
2-polyprenyl-6-methoxy-1,4-benzoquinone methyltransferase activity
rRNA methyltransferase activity
rRNA (uridine-2'-O-)-methyltransferase activity
rRNA (adenine-N6-)-methyltransferase activity
DNA-methyltransferase activity
tRNA (guanine-N1-)-methyltransferase activity
rRNA (cytosine-C5-)-methyltransferase activity
selenocysteine methyltransferase activity
arginine N-methyltransferase activity
protein-arginine N-methyltransferase activity
lysine N-methyltransferase activity
protein-lysine N-methyltransferase activity
tRNA (uracil) methyltransferase activity
tRNA (guanine) methyltransferase activity
tRNA (adenine) methyltransferase activity
tRNA (cytosine) methyltransferase activity
tRNA (cytosine-5-)-methyltransferase activity
tRNA (adenine-N1-)-methyltransferase activity
rRNA (adenine) methyltransferase activity
rRNA (cytosine) methyltransferase activity
rRNA (guanine) methyltransferase activity
rRNA (uridine) methyltransferase activity
transferase activity
protein C-terminal leucine carboxyl O-methyltransferase activity
1-phenanthrol methyltransferase activity
protein-arginine N5-methyltransferase activity
methylarsonite methyltransferase activity
dimethylarsinite methyltransferase activity
4,5-dihydroxybenzo(a)pyrene methyltransferase activity
1-hydroxypyrene methyltransferase activity
1-hydroxy-6-methoxypyrene methyltransferase activity
demethylmenaquinone methyltransferase activity
cobalt-precorrin-6B C5-methyltransferase activity
cobalt-precorrin-7 C15-methyltransferase activity
cobalt-precorrin-5B C1-methyltransferase activity
cobalt-precorrin-3 C17-methyltransferase activity
dimethylamine methyltransferase activity
hydroxyneurosporene-O-methyltransferase activity
tRNA (adenine-57, 58-N(1)-) methyltransferase activity
methylamine-specific methylcobalamin:coenzyme M methyltransferase activity
trimethylamine methyltransferase activity
methanol-specific methylcobalamin:coenzyme M methyltransferase activity
monomethylamine methyltransferase activity
P-methyltransferase activity
Se-methyltransferase activity
2-phytyl-1,4-naphthoquinone methyltransferase activity
tRNA (uracil-2'-O-)-methyltransferase activity
tRNA (cytosine-2'-O-)-methyltransferase activity
phosphomethylethanolamine N-methyltransferase activity
tRNA (cytosine-3-)-methyltransferase activity
rRNA (cytosine-2'-O-)-methyltransferase activity
rRNA (cytosine-N4-)-methyltransferase activity
trihydroxyferuloyl spermidine O-methyltransferase activity
histone-glutamine methyltransferase activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27708643 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27708644 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27708637 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27708641 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27708636 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27708642 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27708645 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27708639 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27708640 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1971139 | BC052068.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1971139 | NM_001004147.3 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P156941 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1971139 | BC052068.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1971139 | NM_001004147.3 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27708649 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27708631 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27708656 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27708662 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27708626 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27708646 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27708627 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27708650 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27708639 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27708633 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27708629 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27708643 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27708659 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27708635 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27708663 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27708632 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27708638 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27708647 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27708641 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27708657 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27708637 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27708645 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P156941 | - | - |
| Whole Mouse Genome G4122A | A_52_P387882 | BC052068.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P387882 | BU701095.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P387882 | NM_001004147.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P387882 | AK140117.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P156941 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09704079 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09704067 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09654823 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09704063 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09704066 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27708639 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27708643 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27708644 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27708640 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27708636 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27708645 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27708642 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27708637 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27708641 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27708624 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27708673 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27708626 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27708631 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27708678 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27708647 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27708633 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27708639 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27708616 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27708675 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27708638 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27708635 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27708632 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27708660 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27708685 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27708649 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27708629 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27708641 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27708643 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27708670 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27708617 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27708619 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27708620 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27708622 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27708627 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27708645 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27708655 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27708684 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27708681 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27708671 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27708637 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27708676 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09349942 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27708650 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09145141 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27708635 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09145142 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27708641 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27708676 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27708618 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27708662 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09175994 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27708632 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27708670 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27708649 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09377971 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27708617 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27708659 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27708663 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09704066 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27708657 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27708638 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09741887 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27708626 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27708633 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27708673 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09011781 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27708616 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27708637 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09704074 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09597073 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27708629 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27708645 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27708665 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09586329 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09672842 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27708624 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27708677 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27708658 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09704076 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27708622 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27708631 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27708675 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10385156 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10385158 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10385157 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1443681_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1459881_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10385155 | - | - |
| Mouse Genome 430 2.0 Array | 1443681_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1459881_at | - | - |
| Mouse Genome 430 2.0 Array | 1443681_at | - | - |
| Mouse Genome 430 2.0 Array | 1459881_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 237730_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 237730_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100023378.mm.1 | NM_001004147.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100023378.mm.1 | NM_001004147.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100023378.mm.1 | TC1100002510.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100023377.mm.1 | NM_001004147.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100023377.mm.1 | NM_001004147.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100023377.mm.1 | TC1100002510.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1459881_at | - | - |
| Mouse Expression Set 430 | 1443681_at | - | - |
| Mouse Expression Set 430 | 1443681_PM_at | - | - |
| Mouse Expression Set 430 | 1459881_PM_at | - | - |
| Murine Genome U74v2 Set | 168173_PM_at | - | - |
| Murine Genome U74v2 Set | 168173_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1443681_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1459881_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1459881_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1443681_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 536293 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1259595 | - | - |

Fbll1
Genomic Map