Assay Details
| Gene Symbol: Itlnb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007552.1 | NP_001007553.1 | 2 - 3 | 124 | 86 | |
| GenBank mRNA | AY217760.1 | - | 2 - 3 | 124 | 86 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Itlnb
Genomic Map