Assay Details
Gene Symbol: Rps15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009091.2 | NP_033117.1 | 2 - 3 | 137 | 117 | |
GenBank mRNA | AK135319.1 | - | 2 - 3 | 111 | 117 | |
AV569793.1 | - | 2 - 3 | 137 | 117 | ||
BC010763.1 | - | 1 - 2 | 103 | 4234356 | 117 | |
BC094409.1 | - | 1 - 2 | 102 | 1179896 | 117 | |
M33330.1 | - | 2 - 3 | 117 | 117 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27282241 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27282239 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27282240 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27282243 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2037524 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P124555 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 279976 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P124555 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2037524 | - | - |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P195997 | BC094409.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P195997 | M33330.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P195997 | NM_009091.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P195997 | AK135319.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P195997 | AK146147.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27282225 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27282230 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27282244 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27282246 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27282240 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27282223 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27282237 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27282233 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27282241 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27282224 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27282228 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27282250 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27282235 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02168701 | - | - |
Whole Mouse Genome G4122A | A_51_P195997 | BC094409.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P195997 | M33330.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P195997 | NM_009091.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P195997 | AK135319.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P195997 | AK146147.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P124555 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P127843 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P117286 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08512453 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08512450 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08354267 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27282243 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27282240 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27282241 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27282239 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282228 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282302 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282289 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282320 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282241 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282286 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282316 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282325 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282305 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282322 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282319 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282314 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282296 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282300 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282238 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282284 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282294 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282280 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282246 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282308 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282307 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282312 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282278 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282310 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282311 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282303 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282324 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282298 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282240 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282233 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27282293 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27282235 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27282246 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27282237 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27282250 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08512450 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27282230 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27282231 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27282233 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27282244 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08512453 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27282240 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27282228 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10541521 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10538630 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10364767 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10364768 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1416089_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10541520 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10538629 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10364766 | - | - |
Mouse Genome 430 2.0 Array | 1416088_PM_a_at | - | - |
Mouse Genome 430 2.0 Array | 1416089_at | - | - |
Mouse Genome 430 2.0 Array | 1416089_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1416088_a_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 20054_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 20054_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1000006455.mm.1 | NM_009091.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000006455.mm.1 | NM_009091.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000006455.mm.1 | TC1000000850.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1000006461.mm.1 | NM_009091.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000006461.mm.1 | NM_009091.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000006461.mm.1 | TC1000000850.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1000006459.mm.1 | NM_009091.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000006459.mm.1 | NM_009091.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000006459.mm.1 | TC1000000850.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1000006458.mm.1 | NM_009091.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000006458.mm.1 | NM_009091.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000006458.mm.1 | TC1000000850.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1000006460.mm.1 | NM_009091.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000006460.mm.1 | NM_009091.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000006460.mm.1 | TC1000000850.mm.1 | MTA TC |
Mouse Expression Set 430 | 1416089_at | - | - |
Mouse Expression Set 430 | 1416088_PM_a_at | - | - |
Mouse Expression Set 430 | 1416088_a_at | - | - |
Mouse Expression Set 430 | 1416089_PM_at | - | - |
Murine Genome U74v2 Set | 100602_PM_at | - | - |
Murine Genome U74v2 Set | 100602_at | - | - |
Mouse Genome 430A 2.0 Array | 1416089_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1416088_PM_a_at | - | - |
Mouse Genome 430A 2.0 Array | 1416089_at | - | - |
Mouse Genome 430A 2.0 Array | 1416088_a_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1416089_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1416089_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 897399 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 580100 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0000580100 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2671517 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2671517 | - | - |
mouse-6 v1.1 (Illumina) | scl0020054.1_111-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 580100 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0020054.1_111-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 580100 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2671517 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2671517 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0020054.1_111-S | - | - |
