Assay Details
| Gene Symbol: Amy2a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001160152.1 | NP_001153624.1 | 3 - 4 | 660 | 140 | |
| Gene Symbol: Amy2a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001160151.1 | NP_001153623.1 | 660 | 140 | ||
| Gene Symbol: Amy2a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001160150.1 | NP_001153622.1 | 660 | 140 | ||
| Gene Symbol: Amy2a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042711.2 | NP_001036176.1 | 522 | 140 | ||
| GenBank mRNA | AK078968.1 | - | 520 | 140 | ||
| AK133526.1 | - | 523 | 140 | |||
| BC094924.1 | - | 510 | 6433956 | 140 | ||
| BC100579.1 | - | 517 | 6433256 | 140 | ||
| V00718.1 | - | 523 | 140 | |||
| X02578.1 | - | 521 | 140 | |||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2318090 | AK007385.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2139402 | NM_001160152.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2139402 | NM_001160151.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2139402 | NM_001160150.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2139402 | BC100579.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2139402 | AK078968.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1996314 | NM_001160152.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1996314 | NM_001160151.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1996314 | NM_001160150.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1996314 | X02578.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1996314 | X02579.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2318090 | AK007385.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2139402 | NM_001160152.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2139402 | NM_001160151.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2139402 | NM_001160150.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2139402 | BC100579.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2139402 | AK078968.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1996314 | NM_001160152.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1996314 | NM_001160151.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1996314 | NM_001160150.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1996314 | X02578.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1996314 | X02579.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P415555 | NM_001160152.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P415555 | NM_001160151.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P415555 | NM_001160150.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P415555 | BC100579.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P415555 | V00718.1 | mouse_transcriptome |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10501497 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10501536 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10501510 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10501534 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10501508 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10501496 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10501498 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10501542 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10501527 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10501501 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10501514 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10501531 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10501520 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10501524 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10501506 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10501499 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10501518 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10501523 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10501537 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10501521 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10501512 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10501509 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10501513 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10501519 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10501503 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10501535 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10501504 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10501541 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10501507 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10501517 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10501528 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10501529 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10501502 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10501543 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10501516 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10501515 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10501539 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10501505 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10501532 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10501540 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10501526 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10501495 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10501525 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10501530 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10501538 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10501494 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10501544 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10501500 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10501522 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10501533 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10501511 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0300021850.mm.1 | NM_001042711.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300021850.mm.1 | NM_001042711.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300021850.mm.1 | TC0300002781.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300021875.mm.1 | NM_001160150.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300021875.mm.1 | NM_001160150.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300021875.mm.1 | TC0300002782.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300021892.mm.1 | NM_001160151.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300021892.mm.1 | NM_001160151.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300021892.mm.1 | NM_001160152.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300021892.mm.1 | TC0300002783.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300021894.mm.1 | NM_001160151.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300021894.mm.1 | NM_001160151.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300021894.mm.1 | NM_001160152.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300021894.mm.1 | TC0300002783.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300021849.mm.1 | NM_001042711.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300021849.mm.1 | NM_001042711.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300021849.mm.1 | TC0300002781.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300021872.mm.1 | NM_001160150.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300021872.mm.1 | NM_001160150.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300021872.mm.1 | TC0300002782.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300021886.mm.1 | NM_001160151.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300021886.mm.1 | NM_001160151.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300021886.mm.1 | NM_001160152.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300021886.mm.1 | TC0300002783.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300021847.mm.1 | NM_001042711.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300021847.mm.1 | NM_001042711.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300021847.mm.1 | TC0300002781.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300021852.mm.1 | NM_001042711.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300021852.mm.1 | NM_001042711.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300021852.mm.1 | TC0300002781.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300021865.mm.1 | NM_001160150.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300021865.mm.1 | NM_001160150.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300021865.mm.1 | TC0300002782.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300021883.mm.1 | NM_001160151.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300021883.mm.1 | NM_001160151.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300021883.mm.1 | NM_001160152.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300021883.mm.1 | TC0300002783.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300021891.mm.1 | NM_001160151.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300021891.mm.1 | NM_001160151.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300021891.mm.1 | NM_001160152.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300021891.mm.1 | TC0300002783.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300021851.mm.1 | NM_001042711.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300021851.mm.1 | NM_001042711.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300021851.mm.1 | TC0300002781.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300021854.mm.1 | NM_001042711.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300021854.mm.1 | NM_001042711.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300021854.mm.1 | TC0300002781.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300021869.mm.1 | NM_001160150.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300021869.mm.1 | NM_001160150.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300021869.mm.1 | TC0300002782.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300021873.mm.1 | NM_001160150.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300021873.mm.1 | NM_001160150.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300021873.mm.1 | TC0300002782.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300021864.mm.1 | NM_001160150.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300021864.mm.1 | NM_001160150.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300021864.mm.1 | TC0300002782.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300021871.mm.1 | NM_001160150.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300021871.mm.1 | NM_001160150.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300021871.mm.1 | TC0300002782.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300021887.mm.1 | NM_001160151.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300021887.mm.1 | NM_001160151.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300021887.mm.1 | NM_001160152.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300021887.mm.1 | TC0300002783.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300021889.mm.1 | NM_001160151.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300021889.mm.1 | NM_001160151.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300021889.mm.1 | NM_001160152.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300021889.mm.1 | TC0300002783.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300021890.mm.1 | NM_001160151.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300021890.mm.1 | NM_001160151.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300021890.mm.1 | NM_001160152.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300021890.mm.1 | TC0300002783.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300021893.mm.1 | NM_001160151.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300021893.mm.1 | NM_001160151.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300021893.mm.1 | NM_001160152.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300021893.mm.1 | TC0300002783.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300021846.mm.1 | NM_001042711.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300021846.mm.1 | NM_001042711.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300021846.mm.1 | TC0300002781.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300021874.mm.1 | NM_001160150.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300021874.mm.1 | NM_001160150.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300021874.mm.1 | TC0300002782.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300021876.mm.1 | NM_001160150.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300021876.mm.1 | NM_001160150.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300021876.mm.1 | TC0300002782.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300021853.mm.1 | NM_001042711.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300021853.mm.1 | NM_001042711.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300021853.mm.1 | TC0300002781.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300021868.mm.1 | NM_001160150.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300021868.mm.1 | NM_001160150.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300021868.mm.1 | TC0300002782.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1416055_PM_at | - | - |
| Mouse Expression Set 430 | 1416055_at | - | - |
| Murine Genome U74v2 Set | 97523_PM_i_at | - | - |
| Murine Genome U74v2 Set | 97524_PM_f_at | - | - |
| Murine Genome U74v2 Set | 97524_f_at | - | - |
| Murine Genome U74v2 Set | 97523_i_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416055_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416055_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 772579 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseRef-8 v2 Expression BeadChip | ILMN_2933478 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2933478 | - | - |

Amy2a2
Genomic Map