Assay Details
| Gene Symbol: Rpl7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011291.5 | NP_035421.2 | 2 - 3 | 247 | 143 | |
| GenBank mRNA | AK017074.1 | - | 2 - 3 | 217 | 143 | |
| AK050656.1 | - | 2 - 3 | 222 | 143 | ||
| AK088291.1 | - | 2 - 3 | 221 | 143 | ||
| AK135460.1 | - | 2 - 3 | 217 | 143 | ||
| AK146219.1 | - | 2 - 3 | 218 | 143 | ||
| AK150943.1 | - | 2 - 3 | 217 | 143 | ||
| AK167124.1 | - | 2 - 3 | 221 | 143 | ||
| AK168137.1 | - | 2 - 3 | 217 | 143 | ||
| AK168227.1 | - | 2 - 3 | 217 | 143 | ||
| BC025909.1 | - | 2 - 3 | 197 | 5027545 | 143 | |
| BC086786.1 | - | 1 - 2 | 193 | 5701071 | 143 | |
| BC096452.1 | - | 2 - 3 | 200 | 3588544 | 143 | |
| M29016.1 | - | 2 - 3 | 216 | 143 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rpl7
Genomic Map