Assay Details
| Gene Symbol: Psma5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011967.3 | NP_036097.1 | 2 - 3 | 443 | 134 | |
| GenBank mRNA | AF019661.1 | - | 2 - 3 | 158 | 134 | |
| AK143468.1 | - | 2 - 3 | 445 | 134 | ||
| AK160530.1 | - | 2 - 3 | 203 | 134 | ||
| AK160550.1 | - | 2 - 3 | 197 | 134 | ||
| AK160741.1 | - | 2 - 3 | 206 | 134 | ||
| AK161680.1 | - | 2 - 3 | 191 | 134 | ||
| BC007181.1 | - | 2 - 3 | 190 | 134 | ||
| BC010709.1 | - | 2 - 3 | 179 | 2648635 | 134 | |
| BC083342.1 | - | 2 - 3 | 176 | 5700963 | 134 | |
| BC108368.1 | - | 2 - 3 | 173 | 3596530 | 134 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Psma5
Genomic Map