Assay Details
| Gene Symbol: Rpl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013762.2 | NP_038790.2 | 6 - 7 | 924 | 70 | |
| GenBank mRNA | AK088993.1 | - | 6 - 7 | 871 | 70 | |
| AK144518.1 | - | 6 - 7 | 871 | 70 | ||
| AK144816.1 | - | 6 - 7 | 871 | 70 | ||
| AK146502.1 | - | 6 - 7 | 871 | 70 | ||
| AK149494.1 | - | 6 - 7 | 871 | 70 | ||
| AK151057.1 | - | 6 - 7 | 871 | 70 | ||
| AK151084.1 | - | 6 - 7 | 871 | 70 | ||
| AK151142.1 | - | 6 - 7 | 871 | 70 | ||
| AK151426.1 | - | 6 - 7 | 871 | 70 | ||
| AK151596.1 | - | 6 - 7 | 871 | 70 | ||
| AK151740.1 | - | 6 - 7 | 871 | 70 | ||
| AK166433.1 | - | 6 - 7 | 867 | 70 | ||
| AK166825.1 | - | 6 - 7 | 871 | 70 | ||
| AK167787.1 | - | 6 - 7 | 871 | 70 | ||
| AK167947.1 | - | 6 - 7 | 871 | 70 | ||
| AK168040.1 | - | 6 - 7 | 871 | 70 | ||
| AK168059.1 | - | 6 - 7 | 870 | 70 | ||
| AK168126.1 | - | 6 - 7 | 871 | 70 | ||
| AK168128.1 | - | 6 - 7 | 871 | 70 | ||
| AK168224.1 | - | 6 - 7 | 871 | 70 | ||
| AK168744.1 | - | 6 - 7 | 869 | 70 | ||
| AK169060.1 | - | 6 - 7 | 871 | 70 | ||
| AK169235.1 | - | 6 - 7 | 871 | 70 | ||
| AK169321.1 | - | 6 - 7 | 871 | 70 | ||
| AK170819.1 | - | 6 - 7 | 871 | 70 | ||
| AK172280.1 | - | 6 - 7 | 871 | 70 | ||
| AK190439.1 | - | 2 - 3 | 247 | 70 | ||
| AK192125.1 | - | 2 - 3 | 182 | 70 | ||
| AK197531.1 | - | 2 - 3 | 247 | 70 | ||
| AK199153.1 | - | 1 - 2 | 160 | 70 | ||
| AK199385.1 | - | 1 - 2 | 158 | 70 | ||
| AK204620.1 | - | 1 - 2 | 173 | 70 | ||
| AK204785.1 | - | 1 - 2 | 166 | 70 | ||
| AK215968.1 | - | 2 - 3 | 247 | 70 | ||
| BC009655.1 | - | 6 - 7 | 867 | 3485069 | 70 | |
| BC083134.1 | - | 5 - 6 | 853 | 6504713 | 70 | |
| BC085495.1 | - | 5 - 6 | 1375 | 70 | ||
| BC094059.1 | - | 5 - 6 | 852 | 3676103 | 70 | |
| BC098497.1 | - | 4 - 5 | 1429 | 70 | ||
| BC158039.1 | - | 6 - 7 | 871 | 9007342 | 70 | |
| U89417.1 | - | 2 - 3 | 299 | 70 | ||
| Y00225.1 | - | 5 - 6 | 853 | 70 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rpl3
Genomic Map