Assay Details
| Gene Symbol: Rpl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013762.2 | NP_038790.2 | 8 - 9 | 1133 | 153 | |
| GenBank mRNA | AK088993.1 | - | 8 - 9 | 1080 | 153 | |
| AK144518.1 | - | 8 - 9 | 1080 | 153 | ||
| AK144816.1 | - | 8 - 9 | 1080 | 153 | ||
| AK146502.1 | - | 8 - 9 | 1080 | 153 | ||
| AK149494.1 | - | 8 - 9 | 1080 | 153 | ||
| AK151084.1 | - | 8 - 9 | 1080 | 153 | ||
| AK151142.1 | - | 8 - 9 | 1080 | 153 | ||
| AK151426.1 | - | 8 - 9 | 1080 | 153 | ||
| AK151596.1 | - | 8 - 9 | 1080 | 153 | ||
| AK151740.1 | - | 8 - 9 | 1080 | 153 | ||
| AK166433.1 | - | 8 - 9 | 1076 | 153 | ||
| AK166825.1 | - | 8 - 9 | 1080 | 153 | ||
| AK167787.1 | - | 8 - 9 | 1080 | 153 | ||
| AK167947.1 | - | 8 - 9 | 1080 | 153 | ||
| AK168040.1 | - | 8 - 9 | 1080 | 153 | ||
| AK168126.1 | - | 8 - 9 | 1080 | 153 | ||
| AK168128.1 | - | 8 - 9 | 1080 | 153 | ||
| AK168224.1 | - | 8 - 9 | 1080 | 153 | ||
| AK168744.1 | - | 8 - 9 | 1078 | 153 | ||
| AK169060.1 | - | 8 - 9 | 1080 | 153 | ||
| AK169235.1 | - | 8 - 9 | 1080 | 153 | ||
| AK169321.1 | - | 8 - 9 | 1080 | 153 | ||
| AK170819.1 | - | 8 - 9 | 1080 | 153 | ||
| AK172280.1 | - | 8 - 9 | 1080 | 153 | ||
| BC009655.1 | - | 8 - 9 | 1076 | 3485069 | 153 | |
| BC083134.1 | - | 7 - 8 | 1062 | 6504713 | 153 | |
| BC085495.1 | - | 7 - 8 | 1584 | 153 | ||
| BC094059.1 | - | 7 - 8 | 1061 | 3676103 | 153 | |
| BC098497.1 | - | 6 - 7 | 1638 | 153 | ||
| BC158039.1 | - | 8 - 9 | 1080 | 9007342 | 153 | |
| BY581903.1 | - | 2 - 3 | 176 | 153 | ||
| Y00225.1 | - | 7 - 8 | 1062 | 153 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rpl3
Genomic Map