Assay Details
| Gene Symbol: Mlxipl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021455.4 | NP_067430.2 | 2 - 3 | 434 | 119 | |
| GenBank mRNA | AF245475.1 | - | 2 - 3 | 412 | 119 | |
| AF245476.1 | - | 2 - 3 | 412 | 119 | ||
| AF245477.1 | - | 3 - 4 | 346 | 119 | ||
| AF245478.1 | - | 2 - 3 | 412 | 119 | ||
| AF245479.1 | - | 2 - 3 | 412 | 119 | ||
| AK143800.1 | - | 2 - 3 | 421 | 119 | ||
| AK143999.1 | - | 2 - 3 | 435 | 119 | ||
| BC064053.1 | - | 2 - 3 | 407 | 119 | ||
| JQ437838.1 | - | 2 - 3 | 280 | 119 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the JQ437838.1 transcripts.
Set Membership: |
|

Mlxipl
Genomic Map