Assay Details
| Gene Symbol: Eef1g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026007.4 | NP_080283.3 | 4 - 5 | 550 | 103 | |
| GenBank mRNA | AF321126.1 | - | 3 - 4 | 393 | 103 | |
| AK007869.1 | - | 4 - 5 | 395 | 103 | ||
| AK007991.1 | - | 4 - 5 | 397 | 103 | ||
| AK011951.1 | - | 3 - 4 | 392 | 103 | ||
| AK011973.1 | - | 4 - 5 | 397 | 103 | ||
| AK012431.1 | - | 4 - 5 | 396 | 103 | ||
| AK014277.1 | - | 4 - 5 | 397 | 103 | ||
| AK050636.1 | - | 4 - 5 | 401 | 103 | ||
| AK081859.1 | - | 4 - 5 | 398 | 103 | ||
| AK145519.1 | - | 4 - 5 | 551 | 103 | ||
| AK145565.1 | - | 4 - 5 | 399 | 103 | ||
| AK147980.1 | - | 4 - 5 | 398 | 103 | ||
| AK159644.1 | - | 4 - 5 | 397 | 103 | ||
| AK159768.1 | - | 4 - 5 | 397 | 103 | ||
| AK166765.1 | - | 4 - 5 | 396 | 103 | ||
| AK167831.1 | - | 4 - 5 | 398 | 103 | ||
| AK168749.1 | - | 4 - 5 | 397 | 103 | ||
| AK192634.1 | - | 1 - 2 | 106 | 103 | ||
| BC023495.1 | - | 3 - 4 | 360 | 5251765 | 103 | |
| BC083071.1 | - | 4 - 5 | 382 | 6329652 | 103 | |
| BC099413.1 | - | 3 - 4 | 378 | 30839877 | 103 | |
| BC117547.1 | - | 3 - 4 | 393 | 40105371 | 103 | |
| BC118522.1 | - | 3 - 4 | 393 | 40105372 | 103 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC023495.1 transcripts.
Set Membership: |
|

Eef1g
Genomic Map