Assay Details
Gene Symbol: Xkr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011874.1 | NP_001011874.1 | 2 - 3 | 1147 | 164 | |
GenBank mRNA | AY534250.1 | - | 2 - 3 | 1147 | 164 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
