Assay Details
| Gene Symbol: Mprip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012027.2 | NP_036157.2 | 7 - 8 | 1227 | 160 | |
| NM_201245.3 | NP_957697.1 | 7 - 8 | 1227 | 160 | ||
| GenBank mRNA | AK044512.1 | - | 7 - 8 | 977 | 160 | |
| AK155630.1 | - | 7 - 8 | 1008 | 160 | ||
| AK158296.1 | - | 7 - 8 | 1216 | 160 | ||
| AK162530.1 | - | 4 - 5 | 558 | 160 | ||
| AK170849.1 | - | 7 - 8 | 1211 | 160 | ||
| AK171089.1 | - | 7 - 8 | 1216 | 160 | ||
| BC039932.1 | - | 6 - 7 | 751 | 160 | ||
| BC049803.1 | - | 7 - 8 | 753 | 160 | ||
| BC063067.1 | - | 7 - 8 | 1190 | 5721396 | 160 | |
| U73200.1 | - | 7 - 8 | 1087 | 160 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mprip
Genomic Map