Assay Details
| Gene Symbol: Srfbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026040.3 | NP_080316.3 | 3 - 4 | 254 | 151 | |
| GenBank mRNA | AK012865.1 | - | 3 - 4 | 256 | 151 | |
| AK141882.1 | - | 248 | 151 | |||
| AK144845.1 | - | 253 | 151 | |||
| AK147692.1 | - | 232 | 151 | |||
| AY611629.1 | - | 3 - 4 | 227 | 151 | ||
| BC024338.1 | - | 231 | 5009612 | 151 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Srfbp1
Genomic Map