Assay Details
| Gene Symbol: Usp22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004143.4 | NP_001004143.2 | 2 - 3 | 484 | 121 | |
| GenBank mRNA | AK034492.1 | - | 2 - 3 | 459 | 121 | |
| AK040187.1 | - | 2 - 3 | 455 | 121 | ||
| AK155071.1 | - | 2 - 3 | 485 | 121 | ||
| AK160992.1 | - | 2 - 3 | 393 | 121 | ||
| AK220368.1 | - | 2 - 3 | 445 | 121 | ||
| BC080737.1 | - | 2 - 3 | 355 | 30431735 | 121 | |
More Information
Updated Mapping Information
Set Membership: |
|

Usp22
Genomic Map