Assay Details
| Gene Symbol: Usp22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004143.4 | NP_001004143.2 | 3 - 4 | 590 | 146 | |
| GenBank mRNA | AK034492.1 | - | 3 - 4 | 565 | 146 | |
| AK040187.1 | - | 3 - 4 | 561 | 146 | ||
| AK155071.1 | - | 3 - 4 | 591 | 146 | ||
| AK160992.1 | - | 3 - 4 | 499 | 146 | ||
| AK220368.1 | - | 3 - 4 | 551 | 146 | ||
| BC080737.1 | - | 3 - 4 | 461 | 30431735 | 146 | |
More Information
Updated Mapping Information
Set Membership: |
|

Usp22
Genomic Map