Assay Details
| Gene Symbol: Usp22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004143.4 | NP_001004143.2 | 7 - 8 | 1118 | 161 | |
| GenBank mRNA | AK034492.1 | - | 7 - 8 | 1093 | 161 | |
| AK084022.1 | - | 3 - 4 | 276 | 161 | ||
| AK155071.1 | - | 7 - 8 | 1119 | 161 | ||
| AK160992.1 | - | 7 - 8 | 1027 | 161 | ||
| AK220368.1 | - | 7 - 8 | 1079 | 161 | ||
| BC052540.1 | - | 3 - 4 | 413 | 161 | ||
| BC080737.1 | - | 7 - 8 | 989 | 30431735 | 161 | |
More Information
Updated Mapping Information
Set Membership: |
|

Usp22
Genomic Map