Assay Details
| Gene Symbol: Usp22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004143.4 | NP_001004143.2 | 8 - 9 | 1274 | 162 | |
| GenBank mRNA | AK034492.1 | - | 8 - 9 | 1249 | 162 | |
| AK084022.1 | - | 4 - 5 | 432 | 162 | ||
| AK155071.1 | - | 8 - 9 | 1275 | 162 | ||
| AK160992.1 | - | 8 - 9 | 1183 | 162 | ||
| AK220368.1 | - | 8 - 9 | 1235 | 162 | ||
| BC052540.1 | - | 4 - 5 | 569 | 162 | ||
| BC080737.1 | - | 8 - 9 | 1145 | 30431735 | 162 | |
More Information
Updated Mapping Information
Set Membership: |
|

Usp22
Genomic Map