Assay Details
| Gene Symbol: Arhgap33os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_036630.1 | 2 - 3 | 202 | 138 | ||
| GenBank mRNA | BC065044.1 | - | 2 - 3 | 202 | 5685982 | 138 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Arhgap33os
Genomic Map