Assay Details
| Gene Symbol: Arhgap33os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_036630.1 | 4 - 5 | 523 | 112 | ||
| GenBank mRNA | BC065044.1 | - | 4 - 5 | 523 | 5685982 | 112 |
More Information
Updated Mapping Information
Set Membership: |
|

Arhgap33os
Genomic Map