Assay Details
| Gene Symbol: Iqsec2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005475.2 | NP_001005475.1 | 9 - 10 | 2331 | 141 | |
| NM_001114664.2 | NP_001108136.1 | 9 - 10 | 3095 | 141 | ||
| GenBank mRNA | AB275281.1 | - | 9 - 10 | 3095 | 141 | |
| AK220345.1 | - | 3 - 4 | 366 | 141 | ||
| CB236789.1 | - | 281 | 141 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Iqsec2
Genomic Map