Assay Details
| Gene Symbol: Mrpl47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029017.2 | NP_083293.1 | 3 - 4 | 329 | 118 | |
| GenBank mRNA | BC029173.1 | - | 309 | 118 | ||
| BC038065.1 | - | 3 - 4 | 300 | 118 | ||
| CX122942.1 | - | 3 - 4 | 308 | 118 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mrpl47
Genomic Map