Assay Details
| Gene Symbol: Cacul1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030197.2 | NP_084473.1 | 6 - 7 | 1215 | 91 | |
| GenBank mRNA | AK036606.1 | - | 6 - 7 | 1099 | 91 | |
| AK147382.1 | - | 6 - 7 | 1218 | 91 | ||
| AK169791.1 | - | 6 - 7 | 1091 | 91 | ||
| AK171596.1 | - | 6 - 7 | 591 | 91 | ||
| BC013270.1 | - | 6 - 7 | 762 | 91 | ||
| BC026363.1 | - | 6 - 7 | 1185 | 4223313 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cacul1
Genomic Map