Assay Details
| Gene Symbol: Senp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163571.1 | NP_001157043.1 | 6 - 7 | 1350 | 144 | |
| NM_030702.4 | NP_109627.3 | 6 - 7 | 1562 | 144 | ||
| GenBank mRNA | AF194031.1 | - | 5 - 6 | 1243 | 144 | |
| AK045071.1 | - | 6 - 7 | 1439 | 144 | ||
| AK088608.1 | - | 6 - 7 | 1351 | 144 | ||
| AY008764.1 | - | 6 - 7 | 1294 | 144 | ||
| BC037014.1 | - | 6 - 7 | 1328 | 5344274 | 144 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Senp3
Genomic Map