Assay Details
| Gene Symbol: Senp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163571.1 | NP_001157043.1 | 8 - 9 | 1565 | 138 | |
| NM_030702.4 | NP_109627.3 | 8 - 9 | 1777 | 138 | ||
| GenBank mRNA | AF194031.1 | - | 7 - 8 | 1458 | 138 | |
| AK045071.1 | - | 8 - 9 | 1654 | 138 | ||
| AK088608.1 | - | 8 - 9 | 1566 | 138 | ||
| AY008764.1 | - | 8 - 9 | 1509 | 138 | ||
| BC037014.1 | - | 8 - 9 | 1543 | 5344274 | 138 | |
More Information
Updated Mapping Information
Set Membership: |
|

Senp3
Genomic Map