Assay Details
| Gene Symbol: Atp6v1b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK085469.1 | - | 1 - 2 | 144 | 117 | |
| BC013168.1 | - | 1 - 2 | 130 | 117 | ||
| BC058584.1 | - | 1 - 2 | 135 | 117 | ||
| BC080796.1 | - | 1 - 2 | 164 | 117 | ||
More Information
Set Membership: |
|

Atp6v1b1
Genomic Map