Assay Details
| Gene Symbol: Krt72 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213728.1 | NP_998893.1 | 7 - 8 | 1336 | 147 | |
More Information
Updated Mapping Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Krt72| Gene Symbol: Krt72 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213728.1 | NP_998893.1 | 7 - 8 | 1336 | 147 | |
Set Membership: |
|