Assay Details
| Gene Symbol: Krt72 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213728.1 | NP_998893.1 | 8 - 9 | 1374 | 122 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2120016 | NM_213728.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 547727 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 910311 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 922987 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 652252 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 613118 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 855877 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 136437 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P148905 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P150992 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P136871 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2120016 | NM_213728.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30493906 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30493892 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30493923 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30493887 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30493888 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30493917 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30493886 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30493915 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30493898 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30493916 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30493910 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30493930 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30493928 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30493932 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30493931 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30493901 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30493938 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30493896 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30493919 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30493889 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30493933 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30493927 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30493925 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30493903 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30493926 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30493936 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30493890 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30493899 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30493907 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30493913 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30493902 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30493895 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30493885 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30493934 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P102563 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12217583 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12217581 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12217565 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12217552 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12217547 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493807 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493802 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493882 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493795 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493777 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493762 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493791 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493946 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493943 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493899 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493879 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493822 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493858 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493898 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493866 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493813 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493927 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493903 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493861 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493896 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493922 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493915 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493932 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493888 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493945 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493874 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493821 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493934 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493827 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493840 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493918 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493963 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493860 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493782 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493849 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493770 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493828 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493780 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493850 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493769 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493926 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493773 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493784 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493968 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493863 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493886 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493864 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493973 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493799 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493836 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493901 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493835 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493910 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493818 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493759 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493793 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493794 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493938 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493965 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493862 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493930 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493838 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493775 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493778 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493833 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493892 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493847 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493789 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493783 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493797 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493865 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493877 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493758 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493810 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493913 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493964 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493811 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493856 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493816 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493890 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493798 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493895 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493936 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493907 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493880 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493805 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493809 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493940 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493906 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493902 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493887 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493917 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493928 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493876 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493845 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493869 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493771 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493786 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493962 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30493935 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30493919 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30493927 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12217562 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12217571 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12217559 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12428300 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30493925 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30493913 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12217560 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30493935 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30493906 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30493864 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30493888 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30493899 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30493807 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30493946 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12217568 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11874652 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30493922 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30493879 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12217550 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12217557 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12142683 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30493809 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30493936 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30493883 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30493847 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30493940 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11987838 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11874653 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30493928 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30493939 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30493849 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30493861 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30493903 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30493920 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30493850 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30493887 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30493877 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12217578 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12217547 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30493876 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30493896 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12376022 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12033595 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12376017 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30493856 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30493934 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30493907 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11874646 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30493874 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12033583 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30493884 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12428286 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30493895 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30493918 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30493865 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30493869 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30493881 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12033589 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12226319 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30493901 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12226322 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10432826 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10432825 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10432830 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10432827 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10432829 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10432823 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10432822 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10432824 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10432828 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1419840_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10432820 | - | - |
| Mouse Genome 430 2.0 Array | 1419840_at | - | - |
| Mouse Genome 430 2.0 Array | 1419840_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 105866_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 105866_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1500018717.mm.1 | NM_213728.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500018717.mm.1 | NM_213728.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500018717.mm.1 | TC1500002272.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1419840_at | - | - |
| Mouse Expression Set 430 | 1419840_PM_at | - | - |
| Murine Genome U74v2 Set | 95528_at | - | - |
| Murine Genome U74v2 Set | 95528_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1419840_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1419840_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1419840_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1419840_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1221678 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1221678 | - | - |

Krt72
Genomic Map