Assay Details
| Gene Symbol: Krt39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213730.2 | NP_998895.2 | 4 - 5 | 857 | 83 | |
| GenBank mRNA | BC119541.1 | - | 4 - 5 | 857 | 83 | |
| BC128563.1 | - | 4 - 5 | 852 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Krt39
Genomic Map